missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
218.00€
Specifications
| Antigen | HIV-1 gp120 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA.Specifications
| HIV-1 gp120 | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Rabbit | |
| Virus | |
| Q6BBS3 | |
| 155971 | |
| Antibody was raised against 16 amino acid synthetic peptide representing the major HIV-1 consensus sequence near the N-terminal region of V1/V2 loop. The immunogen is located within amino acids 80 - 130 of gp120. Amino Acid Squence: EQMHEDIISLWDQLSK | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| env | |
| IgG | |
| Affinity Purified | |
| 120 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title