missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 gp120 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-76371-0.025mg
This item is not returnable.
View return policy
Description
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| HIV-1 gp120 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| Rabbit | |
| 120 kDa | |
| 0.025 mg | |
| Primary | |
| Virus | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000 | |
| Q6BBS3 | |
| env | |
| Antibody was raised against 16 amino acid synthetic peptide representing the major HIV-1 consensus sequence near the N-terminal region of V1/V2 loop. The immunogen is located within amino acids 80 - 130 of gp120. Amino Acid Squence: EQMHEDIISLWDQLSK | |
| Affinity Purified | |
| RUO | |
| 155971 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction