missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Nucleosome Core Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
209.00€ - 371.00€
Specifications
| Antigen | Histone Nucleosome Core |
|---|---|
| Concentration | 2.42 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18262242
|
Novus Biologicals
NB100-745-0.025MG |
0.025 mg |
209.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18767233
|
Novus Biologicals
NB100-745 |
0.125 mg |
371.00€
125µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone Nucleosome Core Polyclonal specifically detects Histone Nucleosome Core in Bovine samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Histone Nucleosome Core | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine | |
| Intact calf H-3 complexed to rRNA. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| 2.42 mg/mL | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Sheep | |
| Chromatin Research, Epigenetics, Histones and Modified Histones | |
| PBS with 0.1% Sodium Azide | |
| IgG | |
| Protein G purified | |
| This is specific for Histone-Core. The antibody binds to H1, H2B, H3 and H4. There is no evidence to suggest that the antibody can bind to H2A. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title