missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone Nucleosome Core Antibody, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-745-0.025MG
This item is not returnable.
View return policy
Description
Histone Nucleosome Core Polyclonal specifically detects Histone Nucleosome Core in Bovine samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Histone Nucleosome Core | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.1% Sodium Azide | |
| Intact calf H-3 complexed to rRNA. | |
| 0.025 mg | |
| Chromatin Research, Epigenetics, Histones and Modified Histones | |
| This is specific for Histone-Core. The antibody binds to H1, H2B, H3 and H4. There is no evidence to suggest that the antibody can bind to H2A. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 2.42 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
| Sheep | |
| Protein G purified | |
| RUO | |
| Primary | |
| Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction