Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Resin
Product Code. p-8010672 Shop All IBA Lifesciences Products
Click to view available options
Quantity:
4 mL
20 mL
50 mL
1000 mL
Unit Size:
1 L
20mL
4mL
50mL
Product Code. 12856456
Brand: IBA Lifesciences 21208002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Strep-Tactin™ Superflow™ High Capacity Resin is a 50% suspension to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
  • Strep-Tactin™ Superflow™ high capacity is a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins.
  • In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
  • The suspension can be used for packing of gravity flow columns since the resin is optimized for column affinity chromatography, rather than for batch purification.
  • To pack a column with 1 ml resin bed volume, 2 ml 50% suspension is necessary.  
Specifications
Quantity 4 mL
Storage Requirements 2°C to 8°C
Binding Capacity Biotin: > 900 nmol/ mL resin
Product Type High Capacity Resin Suspension
For Research Use Only.