missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HGF Antibody (TAK-701) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28396-100ug
This item is not returnable.
View return policy
Description
HGF Monoclonal antibody specifically detects HGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| HGF | |
| Monoclonal | |
| Unconjugated | |
| P14210 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| TAK-701 | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A | |
| HGF/ SF | |
| 100 μg | |
| Cancer, Cell Cycle and Replication | |
| 3082 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction