missing translation for 'onlineSavingsMsg'
Learn More

HDAC3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications

213.00€

Specifications

Antigen HDAC3
Concentration 12.5 mg/mL
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:2500, Immunohistochemistry-Paraffin 1:500, Microarray
Applications Western Blot
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18504614
View Documents
Novus Biologicals
NB500-126-0.025ml
0.025 mL
213.00€
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HDAC3 Polyclonal specifically detects HDAC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
Specifications

Specifications

HDAC3
Western Blot 1:2000-1:10000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:2500, Immunohistochemistry-Paraffin 1:500, Microarray
Polyclonal
Purified
RUO
Human
O15379
8841
Synthetic peptide: C-NEFYDGDHDNDKESDVEI conjugated to KLH, corresponding to amino acids 411-428 of human and chicken HDAC3.
Primary
Store at -20°C. Avoid freeze/thaw cycles.
50 kDa
12.5 mg/mL
Western Blot
Unconjugated
Rabbit
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases
10mM PBS (pH 7.4) with 15mM Sodium Azide
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
HDAC3
IgG
IgG purified
Anti-Histone Deacetylase 3 specifically recognizes histone deacetylase 3 by immunoblotting and immunoprecipitation (50 kD).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.