missing translation for 'onlineSavingsMsg'
Learn More

HDAC3 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 7 publications

Brand:  Novus Biologicals NB500-126-0.025ml

 View more versions of this product

Product Code. 18504614

  • 213.00€ / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HDAC3 Polyclonal specifically detects HDAC3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Microarray.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HDAC3
Polyclonal
Unconjugated
10mM PBS (pH 7.4) with 15mM Sodium Azide
EC 3.5.1.98, HD3RPD3-2RPD3, histone deacetylase 3, SMAP45
Rabbit
50 kDa
0.025 mL
Apoptosis, Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases
8841
Human
Purified
Western Blot
12.5 mg/mL
Western Blot 1:2000-1:10000, Immunohistochemistry 1:500, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 1:2500, Immunohistochemistry-Paraffin 1:500, Microarray
O15379
HDAC3
Synthetic peptide: C-NEFYDGDHDNDKESDVEI conjugated to KLH, corresponding to amino acids 411-428 of human and chicken HDAC3.
IgG purified
RUO
Primary
Anti-Histone Deacetylase 3 specifically recognizes histone deacetylase 3 by immunoblotting and immunoprecipitation (50 kD).
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only