missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HDAC10 Antibody (SR3178), Novus Biologicals™
Rabbit Monoclonal Antibody
327.00€ - 382.00€
Specifications
| Antigen | HDAC10 |
|---|---|
| Clone | SR3178 |
| Dilution | Western Blot 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041558
|
Novus Biologicals™
NBP3-27754-50ul |
50 μL |
327.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041644
|
Novus Biologicals™
NBP3-27754-100ul |
100 μL |
382.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HDAC10 Monoclonal antibody specifically detects HDAC10 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| HDAC10 | |
| Western Blot 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722 | |
| A synthesized peptide derived from human HDAC10 (Uniprot #: Q969S8) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3178 | |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Transcription Factors and Regulators | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 83933 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title