Learn More
Novus Biologicals™ HDAC10 Antibody (SR3178), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27754-50ul
 View more versions of this product
Product Code. 30041558
  • 327.00€ / 50µL
Description
HDAC10 Monoclonal antibody specifically detects HDAC10 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HDAC10
Monoclonal
Unconjugated
PBS (pH 7.4), 150mM NaCl, 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse
Purified
Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
SR3178
Western Blot 1:1000-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
DKFZP761B039, EC 3.5.1.98, HD10, histone deacetylase 10, MGC149722
A synthesized peptide derived from human HDAC10 (Uniprot #: Q969S8)
50 μL
Breast Cancer, Cancer, Checkpoint signaling, Chromatin Research, Epigenetics, Transcription Factors and Regulators
83933
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.