missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCoV-NL63 Human Coronavirus Spike RBD Antibody, R&D Systems™

Mouse Monoclonal Antibody
226.00€ - 480.00€
Specifications
| Antigen | Spike RBD |
|---|---|
| Clone | 1044936 |
| Dilution | Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 3-25 ug/mL |
| Applications | Western Blot, Blocking Assay, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18764287
![]() |
R&D Systems
MAB11037-SP |
25 μg |
226.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18730867
![]() |
R&D Systems
MAB11037-100 |
100 μg |
480.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Spike RBD Monoclonal antibody specifically detects Spike RBD in HCoV-NL63 samples. It is validated for Western Blot, Blocking Assay, ImmunocytochemistrySpecifications
| Spike RBD | |
| Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Human embryonic kidney cell HEK293-derived HCoV-NL63 Spike RBD protein, Ala475-Asp634, Accession # YP_003767.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1044936 | |
| Western Blot, Blocking Assay, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| HCoV-NL63 | |
| YP_003767.1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title