missing translation for 'onlineSavingsMsg'
Learn More

HCoV-NL63 Human Coronavirus Spike RBD Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18764287
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18764287 25 μg 25µg
18730867 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18764287 Supplier R&D Systems Supplier No. MAB11037SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Spike RBD Monoclonal antibody specifically detects Spike RBD in HCoV-NL63 samples. It is validated for Western Blot, Blocking Assay, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Spike RBD
Applications Western Blot, Blocking Assay, Immunocytochemistry
Classification Monoclonal
Clone 1044936
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Blockade of Receptor-ligand Interaction, Immunocytochemistry 3-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
Gene Accession No. YP_003767.1
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived HCoV-NL63 Spike RBD protein, Ala475-Asp634, Accession # YP_003767.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species HCoV-NL63
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.