missing translation for 'onlineSavingsMsg'
Learn More

HBsAg Antibody (HBsAg/7556R), Novus Biologicals™

Rabbit Monoclonal Antibody

291.00€ - 530.00€

Specifications

Antigen HBsAg
Clone HBsAg/7556R
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin : 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18696704
View Documents
Novus Biologicals
NBP3-23794-20ug
20 μg
291.00€
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18693795
View Documents
Novus Biologicals
NBP3-23794-100ug
100 μg
530.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HBsAg Monoclonal antibody specifically detects HBsAg in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

HBsAg
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cell Biology, Virology Bacteria and Parasites
10 mM PBS with 0.05% BSA
944569
IgG κ
Protein A or G purified
HBsAg/7556R
Immunohistochemistry-Paraffin : 1-2 μg/mL
Monoclonal
Purified
RUO
Human
HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S
Recombinant full-length human HBsAg protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.