missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HBsAg Antibody (HBsAg/7556R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-23794-100ug
This item is not returnable.
View return policy
Description
HBsAg Monoclonal antibody specifically detects HBsAg in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HBsAg | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| HBsAg, HBV major surface antigen, HBV surface antigen, Hepatitis B Virus major surface antigen, Hepatitis B Virus Surface Antigen, Major surface antigen, protein S | |
| Recombinant full-length human HBsAg protein | |
| 100 μg | |
| Cell Biology, Virology Bacteria and Parasites | |
| 944569 | |
| Store at 4°C. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| HBsAg/7556R | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction