missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK1 Antibody (G8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
195.00€
Specifications
| Antigen | GRK1 |
|---|---|
| Clone | G8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2 - 3 μg/mL, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18516141
|
Novus Biologicals
NB120-2775-0.025mg |
0.025 mg | |||||||
Description
GRK1 Monoclonal specifically detects GRK1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| GRK1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Protein Kinase, Signal Transduction, Vision | |
| no buffer with 0.05% Sodium Azide | |
| 6011 | |
| Full length human GRK1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| G8 | |
| Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2 - 3 μg/mL, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14 | |
| GRK1 | |
| IgG1 | |
| Protein G purified | |
| Detects G-protein-associated rhodopsin kinase 1a (GRK1a). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title