missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRK1 Antibody (G8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-2775-0.025mg
This item is not returnable.
View return policy
Description
GRK1 Monoclonal specifically detects GRK1 in Human, Mouse, Rat, Bovine, Chicken samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
| GRK1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunocytochemistry/Immunofluorescence 2 - 3 μg/mL, Immunoprecipitation 1:10 - 1:500 | |
| G protein-coupled receptor kinase 1rhodopsin kinase, GPRK1, RHOKEC 2.7.11, RKEC 2.7.11.14 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects G-protein-associated rhodopsin kinase 1a (GRK1a). | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| G8 | |
| Unconjugated | |
| no buffer with 0.05% Sodium Azide | |
| GRK1 | |
| Full length human GRK1. | |
| 0.025 mg | |
| Protein Kinase, Signal Transduction, Vision | |
| 6011 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction