Learn More
GOLM1 Antibody (9C2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26550-100ul
This item is not returnable.
View return policy
Description
GOLM1 Monoclonal antibody specifically detects GOLM1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| GOLM1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA | |
| 9C2 | |
| Western Blot 1:500-1:5000, Flow Cytometry, ELISA | |
| bA379P1.3, FLJ23608, golgi membrane protein 1, Golgi membrane protein GP73, Golgi phosphoprotein 2C9orf155, golgi protein, 73-kD, GOLPH2 FLJ22634, GP73, GP73 chromosome 9 open reading frame 155, PSEC0257 | |
| A synthesized peptide derived from Human GOLM1 [UniProt Q8NBJ4] | |
| 100 μL | |
| Golgi Apparatus Markers, Signal Transduction | |
| 51280 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |