missing translation for 'onlineSavingsMsg'
Learn More

Goat IgG Horseradish Peroxidase-conjugated Antibody, R&D Systems™

Product Code. 18122631
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18122631 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18122631 Supplier R&D Systems Supplier No. HAF019
  • ONLINE EXCLUSIVE PRICE
    244.32€
    217.35€ / 1mL
    Save 26.97€
    11% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chicken Polyclonal Antibody has been used in 13 publications

IgG Polyclonal specifically detects IgG in Goat samples. It is validated for Immunohistochemistry∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate HRP
Dilution Western Blot
Gene Alias Immunoglobulin G, ImmunoglobulinG
Gene Symbols IGHG1
Host Species Chicken
Immunogen Goat IgG
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Test Specificity Detects goat IgG heavy and light chains in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with mouse IgG, rabbit IgG and human IgG is observed.
Target Species Goat
Content And Storage Do not freeze. 6 months from date of receipt, 2 to 8 degreesC as supplied.
Product Type Antibody
Isotype IgY
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.