missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgE Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
150.00€
Specifications
| Antigen | IgE Heavy Chain |
|---|---|
| Concentration | 4.5 mg/mL |
| Applications | Immunoassay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
IgE Heavy Chain Polyclonal antibody specifically detects IgE Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay.Specifications
| IgE Heavy Chain | |
| Immunoassay | |
| Unconjugated | |
| RUO | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| IGHE | |
| IgE | |
| Affinity purified | |
| Based on IEP, this antibody reacts with heavy epsilon chains on human IgE |
| 4.5 mg/mL | |
| Polyclonal | |
| Goat | |
| Human | |
| 3497 | |
| Purified Human IgE | |
| Secondary | |
| Store at 4°C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title