missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Human IgE Heavy Chain Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-74923-0.5mg
This item is not returnable.
View return policy
Description
IgE Heavy Chain Polyclonal antibody specifically detects IgE Heavy Chain in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunodiffusion, Immunoassay.
Specifications
| IgE Heavy Chain | |
| Polyclonal | |
| Unconjugated | |
| IGHE | |
| Purified Human IgE | |
| 0.5 mg | |
| Secondary | |
| Based on IEP, this antibody reacts with heavy epsilon chains on human IgE | |
| Store at 4°C. Do not freeze. |
| Immunoassay | |
| 4.5 mg/mL | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Goat | |
| Affinity purified | |
| RUO | |
| 3497 | |
| Human | |
| IgE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction