missing translation for 'onlineSavingsMsg'
Learn More

Glutathione S-Transferase pi 1/GSTP1 Antibody (8F6), Novus Biologicals™

Rabbit Monoclonal Antibody

343.00€ - 533.00€

Specifications

Antigen Glutathione S-Transferase pi 1/GSTP1
Clone 8F6
Dilution Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043250
View Documents
Novus Biologicals
NBP3-26540-50ul
50 μL
343.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043368
missing translation for 'viewDocuments'
Novus Biologicals
NBP3-26540-100ul
100 μL
533.00€
100µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Glutathione S-Transferase pi 1/GSTP1 Monoclonal antibody specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications

Specifications

Glutathione S-Transferase pi 1/GSTP1
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI
A synthesized peptide derived from Human Glutathione S-Transferase pi 1/GSTP1 [UniProt P09211]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
8F6
Western Blot, ELISA, Immunohistochemistry
Unconjugated
Rabbit
Asthma, Cancer, Immunology, Neuroscience
PBS, pH 7.4, 150mM NaCl and 50% glycerol
2950
IgG
Affinity purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.