Learn More
Glutathione S-Transferase pi 1/GSTP1 Antibody (8F6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26540-100ul
 View more versions of this product
Product Code. 30043368
  • 533.00€ / 100µL
Description
Glutathione S-Transferase pi 1/GSTP1 Monoclonal antibody specifically detects Glutathione S-Transferase pi 1/GSTP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glutathione S-Transferase pi 1/GSTP1
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunohistochemistry
8F6
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200
deafness, X-linked 7, EC 2.5.1.18, FAEES3, fatty acid ethyl ester synthase III, glutathione S-transferase P, glutathione S-transferase pi 1, GST class-pi, GST3DFN7, GSTP, GSTP1-1, PI
A synthesized peptide derived from Human Glutathione S-Transferase pi 1/GSTP1 [UniProt P09211]
100 μL
Asthma, Cancer, Immunology, Neuroscience
2950
Store at -20 to -70C. Avoid freeze-thaw cycles.