missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione Peroxidase 4/GPX4 Antibody (9D4), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | Glutathione Peroxidase 4/GPX4 |
|---|---|
| Clone | 9D4 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043740
|
Novus Biologicals
NBP3-26545-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043082
|
Novus Biologicals
NBP3-26545-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glutathione Peroxidase 4/GPX4 Monoclonal antibody specifically detects Glutathione Peroxidase 4/GPX4 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Glutathione Peroxidase 4/GPX4 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase | |
| A synthesized peptide derived from Human Glutathione Peroxidase 4/GPX4 [UniProt P36969] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9D4 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2879 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title