Learn More
Glutathione Peroxidase 4/GPX4 Antibody (9D4), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26545-100ul
This item is not returnable.
View return policy
Description
Glutathione Peroxidase 4/GPX4 Monoclonal antibody specifically detects Glutathione Peroxidase 4/GPX4 in Human samples. It is validated for ELISA, Immunohistochemistry
Specifications
| Glutathione Peroxidase 4/GPX4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry | |
| 9D4 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| EC 1.11.1, EC 1.11.1.12, Glutathione peroxidase 4, glutathione peroxidase 4 (phospholipid hydroperoxidase), GPx-4, GSHPx-4, MCSP, PHGPxsnGPx, phospholipid hydroperoxidase, phospholipid hydroperoxide glutathione peroxidase, mitochondrial, snPHGPx, sperm nucleus glutathione peroxidase | |
| A synthesized peptide derived from Human Glutathione Peroxidase 4/GPX4 [UniProt P36969] | |
| 100 μL | |
| Cancer, Endocrinology, Signal Transduction | |
| 2879 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |