missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLI2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | GLI2 |
|---|---|
| Concentration | 0.28 mg/mL |
| Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16818804
|
Proteintech
18989-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16808804
|
Proteintech
18989-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes one of four mammalian counterparts to the fruit fly ′dunce′ gene. The encoded protein has 3′,5′-cyclic-AMP phosphodiesterase activity and degrades cAMP, which acts as a signal transduction molecule in multiple cell types. This gene uses different promoters to generate multiple alternatively spliced transcript variants that encode functional proteins.Specifications
| GLI2 | |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P10070, Q0VGT2 | |
| 14633, 2736, 304729 | |
| Peptide | |
| Primary | |
| -20°C | |
| GLI2 |
| 0.28 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| AW546128, GLI Kruppel family member GLI2, Gli2 | |
| Gli2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title