Learn More
GCAP2 Antibody (A1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB120-5422-0.025mg
 View more versions of this product
Product Code. 18564570
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
GCAP2 Monoclonal specifically detects GCAP2 in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GCAP2
Monoclonal
1 mg/mL
Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500
GUCA1B
Bacterial expressed full length GCAP-2.
0.025 mg
Signal Transduction
2979
Bovine
IgG2a
Western Blot
A1
Unconjugated
DKFZp686E1183, GCAP 2, GCAP2guanylate cyclase-activating protein, photoreceptor 2, Guanylate cyclase activator 1B, guanylate cyclase activator 1B (retina), guanylyl cyclase-activating protein 2, GUCA2, RP48
Mouse
Affinity Purified
RUO
Primary
This is specific for GCAP 2 and does not react with other isotypes.
Store at -20°C. Avoid freeze/thaw cycles.