missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCAP2 Antibody (A1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
195.00€
Specifications
| Antigen | GCAP2 |
|---|---|
| Clone | A1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18564570
|
Novus Biologicals
NB120-5422-0.025mg |
0.025 mg | |||||||
Description
GCAP2 Monoclonal specifically detects GCAP2 in Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| GCAP2 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Bovine | |
| 2979 | |
| Bacterial expressed full length GCAP-2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| A1 | |
| Western Blot 1 μg/mL, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Monoclonal | |
| Mouse | |
| Signal Transduction | |
| DKFZp686E1183, GCAP 2, GCAP2guanylate cyclase-activating protein, photoreceptor 2, Guanylate cyclase activator 1B, guanylate cyclase activator 1B (retina), guanylyl cyclase-activating protein 2, GUCA2, RP48 | |
| GUCA1B | |
| IgG2a | |
| Affinity Purified | |
| This is specific for GCAP 2 and does not react with other isotypes. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title