missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gas6 Antibody (RM0084-6J2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
218.00€ - 533.00€
Specifications
| Antigen | Gas6 |
|---|---|
| Clone | RM0084-6J2 |
| Concentration | LYOPH |
| Dilution | Western Blot 1:500-1:1000, ELISA |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18221695
|
Novus Biologicals
NBP1-21724-0.025MG |
0.025 mg |
218.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18210010
|
Novus Biologicals
NBP1-21724 |
0.1 mg |
533.00€
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Gas6 Monoclonal specifically detects Gas6 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
| Gas6 | |
| LYOPH | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| 2621 | |
| Mouse recombinant Gas6 | |
| Primary |
| RM0084-6J2 | |
| Western Blot 1:500-1:1000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| GAS6 | |
| IgG2 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title