missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gas6 Antibody (RM0084-6J2) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-21724-0.025MG
This item is not returnable.
View return policy
Description
Gas6 Monoclonal specifically detects Gas6 in Mouse samples. It is validated for Western Blot, ELISA.
Specifications
| Gas6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:500-1:1000, ELISA | |
| AXLLGAXL stimulatory factor, AXSFAXL receptor tyrosine kinase ligand, DKFZp666G247, FLJ34709, GAS-6, growth arrest-specific 6, growth arrest-specific protein 6 | |
| Rat | |
| Protein G purified | |
| RUO | |
| 2621 | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| RM0084-6J2 | |
| Unconjugated | |
| A 0.2 um filtered solution in PBS with No Preservative | |
| GAS6 | |
| Mouse recombinant Gas6 | |
| 0.025 mg | |
| Primary | |
| Reconstitute with 0.2 ml sterilized PBS to a final concentration of 0.5 mg/ml. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction