missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
277.00€ - 529.00€
Specifications
| Antigen | Galectin-3BP/MAC-2BP/LGALS3BP |
|---|---|
| Concentration | 0.2mg/mL |
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18462681
|
Novus Biologicals
NBP1-89346-25ul |
25 μL |
277.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18795733
|
Novus Biologicals
NBP1-89346 |
529.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Galectin-3BP/MAC-2BP/LGALS3BP | |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 90K, Basement membrane autoantigen p105, BTBD17B, galectin-3-binding protein, L3 antigen, Lectin galactoside-binding soluble 3-binding protein, lectin, galactoside-binding, soluble, 3 binding protein, M2BP, Mac-2 BP, Mac-2-binding protein, MAC2BP, MAC-2-BP, serum protein 90K, Tumor-associated antigen 90K | |
| LGALS3BP | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| 0.2mg/mL | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 3959 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RHERDAGVVCTNETRSTHTLDLSRELSEALGQIFDSQRGCDLSISVNVQGEDALGFCGHTVILTANLEAQALWKEPGSNVTMSVDAECVPMVRDLLRYFYSRRIDITLSSVKCFHKLAS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title