Learn More
Galectin-3BP/MAC-2BP/LGALS3BP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP1-89346
 View more versions of this product
Product Code. 18795733
  • 529.00€ / 0.10mL
Description
Galectin-3BP/MAC-2BP/LGALS3BP Polyclonal specifically detects Galectin-3BP/MAC-2BP/LGALS3BP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Galectin-3BP/MAC-2BP/LGALS3BP
Polyclonal
Rabbit
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry (Paraffin)
LGALS3BP