missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 5 Antibody (S415-24), Novus Biologicals™
Mouse Monoclonal Antibody
174.00€ - 579.00€
Specifications
| Antigen | GABA-AR alpha 5 |
|---|---|
| Clone | S415-24 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284953
|
Novus Biologicals
NBP2-59700 |
100 μg |
579.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18693617
|
Novus Biologicals
NBP2-59700-25ug |
25 μg |
174.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GABA-AR alpha 5 Monoclonal specifically detects GABA-AR alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| GABA-AR alpha 5 | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184 | |
| GABRA5 | |
| IgG1 | |
| Protein G purified | |
| 55 kDa |
| S415-24 | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 2558 | |
| Fusion protein amino acids 368-419 (last cytoplasmic domain before extracellular C-terminus) of human GABA A Receptor Alpha5. | |
| Primary | |
| Detects approx 30kDa. No cross-reactivity against KChIP2, KChIP3, or KChIP4. |
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title