missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-AR alpha 5 Antibody (S415-24), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59700
This item is not returnable.
View return policy
Description
GABA-AR alpha 5 Monoclonal specifically detects GABA-AR alpha 5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| GABA-AR alpha 5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence | |
| GABA(A) receptor subunit alpha-5, gamma-aminobutyric acid (GABA) A receptor, alpha 5, gamma-aminobutyric acid receptor subunit alpha-5, MGC138184 | |
| Mouse | |
| 55 kDa | |
| 100 μg | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| 2558 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S415-24 | |
| Unconjugated | |
| PBS pH7.4, 50% glycerol with 0.1% Sodium Azide | |
| GABRA5 | |
| Fusion protein amino acids 368-419 (last cytoplasmic domain before extracellular C-terminus) of human GABA A Receptor Alpha5. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 30kDa. No cross-reactivity against KChIP2, KChIP3, or KChIP4. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction