missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 3 Antibody (N87/25), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-47613-0.025mg
This item is not returnable.
View return policy
Description
GABA-A R beta 3 Monoclonal specifically detects GABA-A R beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.
Specifications
| GABA-A R beta 3 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1-10 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:1000 | |
| ECA5, GABA(A) receptor beta-3 subunit, GABA(A) receptor subunit beta-3, GABAA receptor beta-3 subunit, GABA-alpha receptor beta-2 subunit, gamma-aminobutyric acid (GABA) A receptor, beta 3, gamma-aminobutyric acid receptor subunit beta-3, MGC9051 | |
| Mouse | |
| 54 kDa | |
| 0.025 mg | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| 2562 | |
| Human | |
| Purified |
| Western Blot | |
| S87-25 | |
| Unconjugated | |
| P63080 | |
| GABRB3 | |
| Fusion protein amino acids 370-433 of mouse GABA-A-R-Beta 3 | |
| Protein G purified | |
| RUO | |
| Primary | |
| Detects approx 55kDa. No cross-reactivity against GABA-A-R-Beta 2 or -Beta1. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction