missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABA-A R beta 3 Antibody (N87/25), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
174.00€
Specifications
| Antigen | GABA-A R beta 3 |
|---|---|
| Clone | S87-25 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-10 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:1000 |
| Applications | Western Blot |
Description
GABA-A R beta 3 Monoclonal specifically detects GABA-A R beta 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Microarray.Specifications
| GABA-A R beta 3 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Neuronal Cell Markers, Neurotransmission | |
| P63080 | |
| 2562 | |
| Fusion protein amino acids 370-433 of mouse GABA-A-R-Beta 3 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 54 kDa |
| S87-25 | |
| Western Blot 1-10 μg/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ECA5, GABA(A) receptor beta-3 subunit, GABA(A) receptor subunit beta-3, GABAA receptor beta-3 subunit, GABA-alpha receptor beta-2 subunit, gamma-aminobutyric acid (GABA) A receptor, beta 3, gamma-aminobutyric acid receptor subunit beta-3, MGC9051 | |
| GABRB3 | |
| IgG1 | |
| Protein G purified | |
| Detects approx 55kDa. No cross-reactivity against GABA-A-R-Beta 2 or -Beta1. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title