missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G3BP1 Rabbit anti-Human, CoraLite 555, Clone: 7F14, Proteintech
Rabbit Recombinant Antibody
145.00€ - 450.00€
Specifications
| Antigen | G3BP1 |
|---|---|
| Clone | 7F14 |
| Applications | Immunofluorescence |
| Conjugate | CoraLite 555 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17781369
|
Proteintech
CL555-80777-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17679916
|
Proteintech
CL555-80777-100UL |
100 μL |
450.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
G3BP1 antibody apecifically detetects G3BP1 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| G3BP1 | |
| Immunofluorescence | |
| Liquid | |
| Human | |
| BC006997 | |
| G3BP1 | |
| IgG | |
| Protein A purification |
| 7F14 | |
| CoraLite 555 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 10146 | |
| G3BP1 fusion protein Ag3728 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title