Learn More
G3BP1 Rabbit anti-Human, CoraLite 555, Clone: 7F14, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech CL555-80777-100UL
This item is not returnable.
View return policy
Description
G3BP1 antibody apecifically detetects G3BP1 antigen in Human Samples. It is validated for Immunofluorescence.Specifications
| G3BP1 | |
| 7F14 | |
| CoraLite 555 | |
| BC006997 | |
| Rabbit | |
| Protein A purification | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| G3BP1 | |
| G3BP1 fusion protein Ag3728 | |
| 100 μL | |
| 10146 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |