missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ FZR1/CDH1 Antibody (Stem Centrx patent anti-Cadherin-1) - Humanized, Novus Biologicals™

Human Monoclonal Antibody

213.00€ - 920.00€

Specifications

Antigen FZR1/CDH1
Clone Stem Centrx patent anti-Cadherin-1
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30046883
View Documents
Novus Biologicals™
NBP3-28089-100ug
100 μg
338.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30044718
View Documents
Novus Biologicals™
NBP3-28089-50ug
50 μg
213.00€
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30045541
View Documents
Novus Biologicals™
NBP3-28089-1mg
1 mg
920.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FZR1/CDH1 Monoclonal antibody specifically detects FZR1/CDH1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications

Specifications

FZR1/CDH1
Flow Cytometry, ELISA, Functional Assay
Unconjugated
Human
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242fizzy-related protein homolog
CDH1/ E-cadherin/ CD324
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Stem Centrx patent anti-Cadherin-1
Monoclonal
Purified
RUO
Human
P12830
51343
IgG1
Protein A purified
Store at -20°C in powder form. Store at -80°C once reconstituted.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.