Learn More
Novus Biologicals™ FZR1/CDH1 Antibody (Stem Centrx patent anti-Cadherin-1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28089-50ug
 View more versions of this product
Product Code. 30044718
  • 213.00€ / 50µg
Description
FZR1/CDH1 Monoclonal antibody specifically detects FZR1/CDH1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FZR1/CDH1
Monoclonal
Unconjugated
P12830
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Stem Centrx patent anti-Cadherin-1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
CDC20C, CDH1, Cdh1/Hct1 homolog, CDH1CDC20-like 1b, fizzy/cell division cycle 20 related 1 (Drosophila), Fzr, FZR2, FZRCDC20-like protein 1, hCDH1, HCDHFYR, KIAA1242fizzy-related protein homolog
CDH1/ E-cadherin/ CD324
50 μg
Cancer, Cell Cycle and Replication, Cellular Markers, Extracellular Matrix, Plasma Membrane Markers, Signal Transduction
51343