missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frk Antibody (4E1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47458-0.025ml
This item is not returnable.
View return policy
Description
Frk Monoclonal specifically detects Frk in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Frk | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| FRK | |
| Purified recombinant fragment of human FRK expressed in E. Coli. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot | |
| 40 | |
| Unconjugated | |
| EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5 protein tyrosine kinase 5, RAKPTK5, tyrosine-protein kinase FRK | |
| Mouse | |
| 58 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication | |
| 2444 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction