missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Frk Antibody (4E1), Novus Biologicals™
Mouse Monoclonal Antibody
217.00€
Specifications
| Antigen | Frk |
|---|---|
| Clone | 40 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1: 500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
Description
Frk Monoclonal specifically detects Frk in Human samples. It is validated for Western Blot, ELISA.Specifications
| Frk | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| 2444 | |
| Purified recombinant fragment of human FRK expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 40 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Mouse | |
| Cell Cycle and Replication | |
| EC 2.7.10, fyn-related kinaseProtein-tyrosine kinase 5, GTKEC 2.7.10.2, Nuclear tyrosine protein kinase RAK, PTK5 protein tyrosine kinase 5, RAKPTK5, tyrosine-protein kinase FRK | |
| FRK | |
| IgG1 | |
| Affinity Purified | |
| 58 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title