missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXO4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77174-0.025mg
This item is not returnable.
View return policy
Description
FOXO4 Polyclonal specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| FOXO4 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4303 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL | |
| NP_005929 | |
| FOXO4 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FOXO4. The immunogen is located within amino acids 30 - 80 of FOXO4. Amino Acid Squence: NQPSEPPEVEPDLGEK | |
| 0.025 mg | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction