missing translation for 'onlineSavingsMsg'
Learn More

FOXO4 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

178.00€

Specifications

Antigen FOXO4
Concentration 1 mg/mL
Dilution Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18793314
View Documents
Novus Biologicals
NBP1-77174-0.025mg
0.025 mg
178.00€
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FOXO4 Polyclonal specifically detects FOXO4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

FOXO4
Western Blot 0.5-1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 μg/mL
Polyclonal
Rabbit
Human, Mouse, Rat
NP_005929
4303
Antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human FOXO4. The immunogen is located within amino acids 30 - 80 of FOXO4. Amino Acid Squence: NQPSEPPEVEPDLGEK
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
PBS with 0.02% Sodium Azide
AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7
FOXO4
IgG
Affinity Purified
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.