Learn More
Florfenicol, 98%, Thermo Scientific™
217.00€ - 660.00€
Chemical Identifiers
| CAS | 73231-34-2 |
|---|---|
| Molecular Formula | C12H14Cl2FNO4S |
| Molecular Weight (g/mol) | 358.205 |
| MDL Number | MFCD00864834 |
| InChI Key | AYIRNRDRBQJXIF-NXEZZACHSA-N |
| PubChem CID | 114811 |
| ChEBI | CHEBI:87185 |
| IUPAC Name | 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
| SMILES | CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15498579
|
Thermo Scientific Alfa Aesar
J66995.03 |
1g |
217.00€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15408589
|
Thermo Scientific Alfa Aesar
J66995.06 |
5g |
660.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
Chemische Identifikatoren
| 73231-34-2 | |
| 358.205 | |
| AYIRNRDRBQJXIF-NXEZZACHSA-N | |
| CHEBI:87185 | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O |
| C12H14Cl2FNO4S | |
| MFCD00864834 | |
| 114811 | |
| 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide |
Spezifikation
| 73231-34-2 | |
| >110°C (230°F) | |
| MFCD00864834 | |
| 14,4109 | |
| AYIRNRDRBQJXIF-NXEZZACHSA-N | |
| 2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide | |
| 114811 | |
| 358.21 | |
| Florfenicol |
| White | |
| C12H14Cl2FNO4S | |
| 618°C | |
| Soluble in ethanol to 25mM and in DMSO to 100mM | |
| CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O | |
| 358.205 | |
| CHEBI:87185 | |
| 98% |
Sicherheit und Handhabung
missing translation for 'rtecsNumber' : DB6034000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.