missing translation for 'onlineSavingsMsg'
Learn More

Florfenicol, 98%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar J66995.06

Quantity 5g

 View more versions of this product

Product Code. 15408589

  • 660.00€ / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • A bacteriostatic antibiotic similar to chloramphenicol which inhibits bacterial protein synthesis
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

73231-34-2
358.205
AYIRNRDRBQJXIF-NXEZZACHSA-N
CHEBI:87185
CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O
C12H14Cl2FNO4S
MFCD00864834
114811
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide

Specifications

73231-34-2
5g
C12H14Cl2FNO4S
618°C
Soluble in ethanol to 25mM and in DMSO to 100mM
CS(=O)(=O)C1=CC=C(C=C1)C(C(CF)NC(=O)C(Cl)Cl)O
358.205
CHEBI:87185
98%
White
>110°C (230°F)
MFCD00864834
14,4109
AYIRNRDRBQJXIF-NXEZZACHSA-N
2,2-dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulfonylphenyl)propan-2-yl]acetamide
114811
358.21
Florfenicol
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.