missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
218.00€
Specifications
| Antigen | FKBP12 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
FKBP12 Polyclonal specifically detects FKBP12 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| FKBP12 | |
| Western Blot 1 μg/mL, Immunohistochemistry 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Autophagy, Cancer, mTOR Pathway | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| FKBP1A | |
| IgG | |
| Affinity Purified | |
| Detects FK506 binding protein 12 kDa (FKBP12). |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P62942 | |
| 2280 | |
| Synthetic peptide corresponding to the N-terminal residues G(1) V Q V E T I S P G D G R(13) of human FKBP12. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title