missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB300-508-0.025ml
This item is not returnable.
View return policy
Description
FKBP12 Polyclonal specifically detects FKBP12 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| FKBP12 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| 12 kDa FKBP, EC 5.2.1.8, FK506 binding protein 1A, 12kDa, FK506-binding protein 1A, FKBP1, FKBP12C, FKBP12FKBP12-Exip3, FKBP-12T-cell, 12-kD, FKBP-1A, peptidyl-prolyl cis-trans isomerase FKBP1A, PPIase FKBP1A, protein kinase C inhibitor 2, rotamase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Detects FK506 binding protein 12 kDa (FKBP12). | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1 μg/mL, Immunohistochemistry 2 μg/mL, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| P62942 | |
| FKBP1A | |
| Synthetic peptide corresponding to the N-terminal residues G(1) V Q V E T I S P G D G R(13) of human FKBP12. | |
| 0.025 mL | |
| Autophagy, Cancer, mTOR Pathway | |
| 2280 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction