missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen beta chain Antibody (1F9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-47442-0.025ml
This item is not returnable.
View return policy
Description
Fibrinogen beta chain Monoclonal specifically detects Fibrinogen beta chain in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Fibrinogen beta chain | |
| Monoclonal | |
| Unconjugated | |
| Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p, MGC104327, MGC120405 | |
| Mouse | |
| 52 kDa | |
| 0.025 mL | |
| Cell Cycle and Replication | |
| 2244 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot | |
| 1F9 | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| FGB | |
| Purified recombinant fragment of human Fibrinogen beta chain (aa30-300) expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction