missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibrinogen beta chain Antibody (1F9), Novus Biologicals™
Mouse Monoclonal Antibody
203.00€
Specifications
| Antigen | Fibrinogen beta chain |
|---|---|
| Clone | 1F9 |
| Dilution | Western Blot 1: 500-1:2000, ELISA 1:10000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Fibrinogen beta chain Monoclonal specifically detects Fibrinogen beta chain in Human samples. It is validated for Western Blot, ELISA.Specifications
| Fibrinogen beta chain | |
| Western Blot 1: 500-1:2000, ELISA 1:10000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 2244 | |
| Purified recombinant fragment of human Fibrinogen beta chain (aa30-300) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1F9 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication | |
| Beta-Fibrinogen, epididymis secretory sperm binding protein Li 78p, fibrinogen beta chain, fibrinogen, B beta polypeptide, HEL-S-78p, MGC104327, MGC120405 | |
| FGB | |
| IgG1 | |
| Unpurified | |
| 52 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title