missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR3 Antibody (6D4), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00€ - 533.00€
Specifications
| Antigen | FGFR3 |
|---|---|
| Clone | 6D4 |
| Dilution | Western Blot 1:500-1:5000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043202
|
Novus Biologicals
NBP3-26582-50ul |
50 μL |
343.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043238
|
Novus Biologicals
NBP3-26582-100ul |
100 μL |
533.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FGFR3 Monoclonal antibody specifically detects FGFR3 in Human samples. It is validated for Western Blot, ELISASpecifications
| FGFR3 | |
| Western Blot 1:500-1:5000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.10, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4thanatophoric dwarfism | |
| A synthesized peptide derived from Human FGFR3 [UniProt P22607] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6D4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Protein Kinase | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 2261 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title